Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium hydroxide octahydrate ≥98.0% ACS

Supplier: BeanTown Chemical

121485-250G 121485-1KG
BT121485-250GEA 51.9 USD InStock
BT121485-250G BT121485-1KG
Barium hydroxide octahydrate ≥98.0% ACS
Barium hydroxide octahydrate
  • RTECS: CQ9200000
  • Air Sensitive, Hygroscopic
  • Crystalline


Formula: Ba(OH)₂·8H₂O
MW: 315.47 g/mol
Melting Pt: 78 °C
MDL Number: MFCD00149152
CAS Number: 12230-71-6
EINECS: 241-234-5
UN: 3262
ADR: 8,III
Order Now
Specification Test Results
Assay 98.0% min.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty