Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tri-Sodium citrate dihydrate ≥99%

Supplier: BeanTown Chemical
Synonyms: Sodium Citrate, DihydrateCitric acid trisodium salt dihydrateSodium Citrate DihydrateSodium citrate tribasic dihydrateTrisodium citrateSodium citrate dihydrateCitric acid tridosium salt dihydrate

123215-2.5KG 123215-500G
BT123215-2.5KGEA 117.54 USD InStock
BT123215-2.5KG BT123215-500G
tri-Sodium citrate dihydrate ≥99%
tri-Sodium citrate dihydrate
  • Crystalline/Powder


Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Melting Pt: 300 °C
MDL Number: MFCD00150031
CAS Number: 6132-04-3
EINECS: 200-675-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/03/16640603.jpg
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty