Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cobalt(II) chloride hexahydrate ≥99.995% (trace metals basis)

Supplier: BeanTown Chemical
Synonyms: Cobalt dichloride hexahydrateCobalt Chloride, 6-HydrateCobaltous chloride hexahydrateCobalt Chloride, Hexahydrate

123930-10G 123930-50G
BT123930-10GEA 101.9 USD InStock
BT123930-10G BT123930-50G
Cobalt(II) chloride hexahydrate ≥99.995% (trace metals basis)
Cobalt (II) chloride hexahydrate
  • RTECS: GG0200000
  • Hygroscopic
  • Crystalline Aggregates


Formula: CoCl₂·6H₂O
MW: 237.93 g/mol
MDL Number: MFCD00149652
CAS Number: 7791-13-1
EINECS: 231-589-4
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Assay 99.995%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty