Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methanol absolute ≥99.8% ACS low acid

Supplier: BeanTown Chemical
Danger
Synonyms: Hydroxy methaneMethyl hydroxideMethyl alcoholMethylolCarbinolWood alcohol

124220-4L 124220-1L 124220-500ML
BT124220-4LEA 83.99 USD InStock
BT124220-4L BT124220-500ML BT124220-1L
Methanol absolute ≥99.8% ACS low acid
Methanol
  • RTECS: PC1400000
  • Refractive index: 1.329
  • Liquid


Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.7 °C
Melting Pt: –98 °C
Density: 0.791 g/mL
Flash Pt: 9.7 °C (49.5 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg
Order Now
Specification Test Results
Assay 99.8% min.
Color (APHA) ≤10
Residue after evaporation ≤0.001%
Identification, IR Passes test
Identification, GC Passes test
Acidity ≤0.45ml, 0.02N NaOH
Alkalinity (as ammonia) ≤0.20ml, 0.02N H2SO4
Titrable acid ≤0.0003meq/g
Titrable base ≤0.0002meq/g
Readily carbonizable substances Passes test
Acetone & aldehydes ≤0.003%
Readily oxidizable substances Passes test
Non-volatile residue ≤2mg (0.001% w/w)
Water ≤0.10%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty