Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2',7'-Dichlorofluorescein

Supplier: BeanTown Chemical
Synonyms: 2',7'-Dichloro-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

127805-5G 127805-25G
BT127805-5GEA 61.58 USD InStock
BT127805-5G BT127805-25G
2',7'-Dichlorofluorescein
2',7'-Dichlorofluorescein
  • Solid


Formula: C₂₀H₁₀Cl₂O₅
MW: 401.20 g/mol
Melting Pt: 280 °C (dec.)
MDL Number: MFCD00005047
CAS Number: 76-54-0
EINECS: 200-968-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty