Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl acetate ≥99.5% for HPLC

Supplier: BeanTown Chemical
Synonyms: EtOAcAcetidinEthyl ethanoateVinegar naphthaEthyl acetic esterAcetic acid ethyl esterAcetic etherAcetoxyethane

132480-4L 132480-4X1L 132480-1L
BT132480-4X1LEA 227.43 USD InStock
BT132480-4X1L BT132480-4L BT132480-1L
Ethyl acetate ≥99.5% for HPLC
Ethyl acetate
  • RTECS: AH5425000
  • Refractive index: 1.372
  • Liquid


Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 76.5…77.5 °C
Melting Pt: –84 °C
Density: 0.902 g/mL
Flash Pt: –3 °C (27 °F)
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/20/72/722072.jpg
Order Now
Specification Test Results
Assay 99.5% min.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty