Synonyms:
Oxolane
, THF
, Cyclotetramethylene oxide
, Tetramethylene oxide
, 1,4-epoxy-Butane
, Butane alpha,delta-oxide
, Oxacyclopentane
132850-500ML
132850-2.5L
132850-1L
BT132850-2.5LEA
121.37
USD
InStock
BT132850-2.5L
BT132850-500ML
BT132850-1L
Tetrahydrofuran ≥99% stabilized
Tetrahydrofuran
- RTECS: LU5950000
- Refractive index: 1.407
- Liquid