Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lactic acid ≥85.0-90.0% (w/w) in water ACS

Supplier: BeanTown Chemical
Synonyms: 2-Hydroxypropionic acid2-Hydroxypropanoic acidDL-Lactic acid

133595-2KG 133595-500G 133595-100G
BT133595-2KGEA 209.09 USD InStock
BT133595-2KG BT133595-500G BT133595-100G
Lactic acid ≥85.0-90.0% (w/w) in water ACS
Lactic acid
  • RTECS: OD2800000
  • Refractive index: 1.425
  • Liquid


Formula: H₃CCH(OH)COOH
MW: 90.08 g/mol
Density: 1.209 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00004520
CAS Number: 50-21-5
EINECS: 200-018-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty