Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitromethane ≥98%

Supplier: BeanTown Chemical

135205-500G 135205-2.5KG 135205-100G
BT135205-100GEA 35.1 USD InStock
BT135205-100G BT135205-500G BT135205-2.5KG
Nitromethane ≥98%
Nitromethane
  • RTECS: PA9800000
  • Refractive index: 1.382
  • Liquid


Formula: CH₃NO₂
MW: 61.04 g/mol
Boiling Pt: 101 °C
Melting Pt: –29 °C
Density: 1.127 g/mL
Flash Pt: 35 °C (95 °F)
MDL Number: MFCD00007400
CAS Number: 75-52-5
EINECS: 200-876-6
UN: 1261
ADR: 3,II
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty