Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Buffer Solutions, Ready-to- Use, NIST Traceable, BeanTown Chemical

Supplier: BeanTown Chemical

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

220710-500ML 127620-500ML 142055-500ML 142055-4L 127650-4L 142055-1L 220935-4L 220935-1L 127485-4L 220710-1L 220935-500ML 127620-1L 135685-500ML 135685-1L 127650-1L 144730-4L 144755-1L 144755-4L 144755-500ML 127650-500ML 222065-1L 144730-1L 127485-1L 127485-500ML 144730-500ML 222065-4L 222065-500ML 135685-4L 127620-4L 220710-4L
BT220710-1LEA 34.59 USD InStock
BT220710-1L BT144755-500ML BT127620-1L BT135685-500ML BT144730-500ML BT222065-4L BT135685-1L BT220935-4L BT144755-1L BT127650-500ML BT220935-500ML BT127650-1L BT144730-1L BT142055-4L BT127485-4L BT222065-1L BT127620-4L BT127620-500ML BT127485-500ML BT144755-4L BT220710-500ML BT135685-4L BT220935-1L BT142055-500ML BT127485-1L BT127650-4L BT144730-4L BT220710-4L BT142055-1L BT222065-500ML
Buffer Solutions, Ready-to- Use, NIST Traceable, BeanTown Chemical
Buffers pH Calibration Buffers
MDL No: MFCD00147253

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty