Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 0.5 N in water volumetric solution

Supplier: BeanTown Chemical
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

137880-4L 137880-1L 137880-20L
BT137880-20LEA 137.96 USD InStock
BT137880-20L BT137880-1L BT137880-4L
Sodium hydroxide 0.5 N in water volumetric solution
Sodium hydroxide
  • Air Sensitive
  • Liquid


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 105…140 °C
Melting Pt: –12–10 °C
Density: 1.02 g/mL
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 3266
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty