Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

N-Methyl-2-pyrrolidone (NMP) ≥99.5% for HPLC

Supplier: BeanTown Chemical
Synonyms: N-Methyl-alpha-pyrrolidone1-Methyl-2-pyrrolidoneNMPN-Methyl-2-pyrrolidoneN-Methylpyrrolidone1-Methyl-2-pyrrolidinoneN-MethylpyrrolidinoneN-methylpyrrolidoneN-Methyl-gamma-butyrolactamN-Methyl-2-pyrrolidinoneM-Pyrol

138465-1L 138465-4X1L 138465-4L
BT138465-1LEA 121.46 USD InStock
BT138465-1L BT138465-4L BT138465-4X1L
N-Methyl-2-pyrrolidone (NMP) ≥99.5% for HPLC
N-Methyl-2-pyrrolidone (NMP)
  • RTECS: UY5790000
  • Refractive index: 1.470
  • Liquid


Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202 °C
Melting Pt: –24 °C
Density: 1.028 g/mL
Flash Pt: 91 °C (196 °F)
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/88/54/2298854.jpg
Order Now
Specification Test Results
Assay 99.5% min.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty