Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Strontium, Plasma Standard Solution, Sr 1000 µg/ml, BeanTown Chemical
Supplier: BeanTown Chemical
139360-100ML 139360-500ML 139360-50ML
BT139360-500MLEA 221.31 USD InStock
BT139360-500ML BT139360-50ML BT139360-100ML
Strontium, Plasma Standard Solution, Sr 1000 µg/ml, BeanTown Chemical
Standards Cationic Standards Strontium Standards
EC No: 231-714-2; MDL No: MFCD00134060
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty