Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrabutylammonium iodide ≥98%

Supplier: BeanTown Chemical
Synonyms: TBAITetra-n-butylammonium iodide

139565-250G 139565-1KG 139565-50G
BT139565-1KGEA 347.96 USD InStock
BT139565-1KG BT139565-50G BT139565-250G
Tetrabutylammonium iodide ≥98%
Tetrabutylammonium iodide
  • RTECS: BS5450000
  • Hygroscopic, Light Sensitive
  • Crystalline/Powder


Formula: C₁₆H₃₆IN
MW: 369.37 g/mol
Melting Pt: 141…143 °C
MDL Number: MFCD00011636
CAS Number: 311-28-4
EINECS: 206-220-5
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty