Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,1,1-Trichloro-2-methyl-2-propanol hemihydrate ≥98%

Supplier: BeanTown Chemical
Synonyms: Chlorbutanol hemihydrateChlorobutanol hydrous1,1,1-Trichloro-2-methyl-2-propanol Hemihydrate

140390-100G 140390-25G
BT140390-100GEA 89.54 USD InStock
BT140390-100G BT140390-25G
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate ≥98%
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate
  • RTECS: UC0175000
  • Solid


Formula: C₈H₁₆Cl₆O₃
MW: 372,93 g/mol
Melting Pt: 77…79 °C
Flash Pt: 100 °C (212 °F)
MDL Number: MFCD00004461
CAS Number: 6001-64-5
EINECS: 200-317-6
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty