Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphorus amorphous, red ≥99% (trace metals basis), powder -100 mesh

Supplier: BeanTown Chemical

141055-4X500G 141055-100G 141055-500G
BT141055-4X500EA 399.89 USD InStock
BT141055-4X500 BT141055-500G BT141055-100G
Phosphorus amorphous, red ≥99% (trace metals basis), powder -100 mesh
Phosphorus
  • RTECS: TH3495000
  • Powder


Formula: P
MW: 30.97 g/mol
Melting Pt: 416 °C (subl.)
Density: 2.34 g/mL
MDL Number: MFCD00133771
CAS Number: 7723-14-0
EINECS: 231-768-7
UN: 1338
ADR: 4.1,III
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty