Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide ≥98%, flakes

Supplier: BeanTown Chemical
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

142245-500G 142245-2.5KG 142245-10KG
BT142245-2.5KGEA 95.71 USD InStock
BT142245-2.5KG BT142245-10KG BT142245-500G
Sodium hydroxide ≥98%, flakes
Sodium hydroxide
  • RTECS: WB4900000
  • Air Sensitive, Hygroscopic
  • Flakes


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C
Melting Pt: 318 °C
Density: 2.13 g/mL
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty