Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium nitrite ≥99.999% (trace metals basis)

Supplier: BeanTown Chemical

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger

142445-100G 142445-25G
BT142445-100GEA 327.81 USD InStock
BT142445-100G BT142445-25G
Sodium nitrite ≥99.999% (trace metals basis)
Sodium nitrite
  • RTECS: RA1225000
  • Hygroscopic


Formula: NaNO₂
MW: 69 g/mol
Boiling Pt: 320 °C
Melting Pt: 271 °C
Density: 2.168
MDL Number: MFCD00011118
CAS Number: 7632-00-0
EINECS: 231-555-9
UN: 1500
ADR: 5.1 (6.1),III
Order Now
Specification Test Results
Assay 99.999%

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty