Would you like to log in again?
Selenium, AAS standard solution, Se 1000 µg/ml, BeanTown Chemical
Supplier: BeanTown Chemical
143130-500ML 143130-100ML
BT143130-100MLEA 40.59 USD InStock
BT143130-100ML BT143130-500ML
Selenium, AAS standard solution, Se 1000 µg/ml, BeanTown Chemical
Standards Cationic Standards Selenium Standards
EC No: 231-714-2; MDL No: MFCD00134090
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty