Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

cis-Platin

Supplier: BeanTown Chemical
Synonyms: cis-Diaminodichloroplatinum(II)cis-Diamminedichloroplatinum (II)cis-Diamineplatinum(II) dichloridePlatinolcis-Dichlorodiammine platinum(II)Cisplatin

143840-5G 143840-1G 143840-250MG
BT143840-250MGEA 83.42 USD InStock
BT143840-250MG BT143840-1G BT143840-5G
cis-Platin
cis-Platin
  • RTECS: TP2450000
  • Crystalline


Formula: H₆Cl₂N₂Pt
MW: 300.05 g/mol
Melting Pt: 270 °C (dec.)
MDL Number: MFCD00011623
CAS Number: 15663-27-1
EINECS: 239-733-8
UN: 3288
ADR: 6.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty