Synonyms:
(S)-(-)-α-Terpineol
, 2-(4-Methylcyclohex-3-en-1-yl)propan-2-ol
144440-500G
144440-25G
144440-100G
BT144440-100GEA
40.3
USD
InStock
BT144440-100G
BT144440-500G
BT144440-25G
(±)-α-Terpineol ≥96%
(±)-α-Terpineol
- RTECS: WZ6700000
- Refractive index: 1.482
- Liquid