Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrahydrofuran ≥99.7% stabilized, non-UV for HPLC

Supplier: BeanTown Chemical
Synonyms: OxolaneTHFCyclotetramethylene oxideTetramethylene oxide1,4-epoxy-ButaneButane alpha,delta-oxideOxacyclopentane

144445-1L 144445-4L 144445-4X1L
BT144445-4LEA 307.13 USD InStock
BT144445-4L BT144445-4X1L BT144445-1L
Tetrahydrofuran ≥99.7% stabilized, non-UV for HPLC
Tetrahydrofuran
  • RTECS: LU5950000
  • Refractive index: 1.407
  • Liquid


Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 65…67 °C
Melting Pt: –108 °C
Density: 0.889 g/mL
Flash Pt: –17 °C (1 °F)
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/95/19/789519.jpg
Order Now
Specification Test Results
Assay 99.7% min.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty