Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Perchloric acid 0.1 N in acetic acid volumetric solution

Supplier: BeanTown Chemical

145850-4L 145850-1L
BT145850-4LEA 303.18 USD InStock
BT145850-4L BT145850-1L
Perchloric acid 0.1 N in acetic acid volumetric solution
Perchloric acid
  • Liquid


Formula: HClO₄
MW: 100.46 g/mol
Boiling Pt: 120 °C
Density: 1.05 g/mL
Flash Pt: 40 °C (104 °F)
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 2920
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/22/16640622.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty