Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 4 M in 1,4-dioxane ≥99%

Supplier: BeanTown Chemical
Synonyms: Muriatic acid

212670-100ML 212670-50ML
BT212670-50MLEA 63.33 USD InStock
BT212670-50ML BT212670-100ML
Hydrochloric acid 4 M in 1,4-dioxane ≥99%
Hydrochloric acid
  • Hygroscopic
  • Liquid


Formula: HCl
MW: 36.46 g/mol
Density: 1.05 g/mL
Flash Pt: 17 °C (63 °F)
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 2924
ADR: 8,II
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty