Would you like to log in again?
Calcium Carbonate, Chelometric Standard, ACS, 99.95-100.05%
Supplier: BeanTown Chemical
213950-500G 213950-100G
BT213950-100GEA 68.41 USD InStock
BT213950-100G BT213950-500G
Calcium Carbonate, Chelometric Standard, ACS, 99.95-100.05%
Standards Cationic Standards Calcium Standards
May be used as a chelometric standard for the standardization of EDTA solution.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty