Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide ≥98%, pellets

Supplier: BeanTown Chemical
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

214430-500G
BT214430-500GEA 28.86 USD InStock
BT214430-500G
Sodium hydroxide ≥98%, pellets
Sodium hydroxide
  • RTECS: WB4900000
  • Air Sensitive, Hygroscopic
  • Pellets


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C
Melting Pt: 318 °C
Density: 2.13 g/mL
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty