Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 10.0 N in water volumetric solution

Supplier: BeanTown Chemical
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

214540-1L
BT214540-1LEA 46.16 USD InStock
BT214540-1L
Sodium hydroxide 10.0 N in water volumetric solution
Sodium hydroxide
  • Air Sensitive
  • Liquid


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 105…140 °C
Melting Pt: –12–10 °C
Density: 1.327 g/mL
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty