Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphorus(V) chloride ≥98%

Supplier: BeanTown Chemical

215220-500G 215220-100G
BT215220-100GEA 47.6 USD InStock
BT215220-100G BT215220-500G
Phosphorus(V) chloride ≥98%
Phosphorus (V) chloride
  • RTECS: TB6125000
  • Moisture Sensitive
  • Powder


Formula: Cl₅P
MW: 208.24 g/mol
Melting Pt: 160 °C (subl.)
Density: 2.114 g/mL
MDL Number: MFCD00011439
CAS Number: 10026-13-8
EINECS: 233-060-3
UN: 1806
ADR: 8,II
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty