Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl ethyl ketone ≥99%

Supplier: BeanTown Chemical
Synonyms: 2-ButanoneMEKButan-2-oneEK 500 AdditiveButanoneEthyl methyl ketone

217280-2.5L 217280-500ML 217280-1L
BT217280-500MLEA 35.63 USD InStock
BT217280-500ML BT217280-2.5L BT217280-1L
Methyl ethyl ketone ≥99%
Methyl ethyl ketone
  • RTECS: EL6475000
  • Refractive index: 1.379
  • Liquid


Formula: C₂H₅COCH₃
MW: 72.11 g/mol
Boiling Pt: 80 °C
Melting Pt: –87 °C
Density: 0.805 g/mL
Flash Pt: –3 °C (27 °F)
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/03/79/720379.jpg
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty