Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 36.5 - 38.0% ACS

Supplier: BeanTown Chemical
Synonyms: Muriatic acid

217905-6X6LB 217905-6LB
BT217905-6X6LBEA 349.01 USD InStock
BT217905-6X6LB BT217905-6LB
Hydrochloric acid 36.5 - 38.0% ACS
Hydrochloric acid
  • RTECS: MW4025000
  • Liquid


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: >100 °C
Density: 1.19 g/mL
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Assay 36.5-38.0%
Color (APHA) ≤10
Extractable organic substances ≤5ppm
Arsenic (As) ≤0.01ppm
Ammonium (NH4) ≤3ppm
Bromide (Br) ≤0.005%
Free chloride (Cl) ≤1ppm
Heavy metals (as Pb) ≤1ppm
Iron (Fe) ≤0.2ppm
Residue after ignition ≤5ppm
Sulfate (SO4) ≤1ppm
Sulfite (SO3) ≤1ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty