Would you like to log in again?
Zirconium, Plasma Standard Solution, Zr 1000 µg/ml, BeanTown Chemical
Supplier: BeanTown Chemical
219075-100ML 219075-50ML 219075-500ML
BT219075-500MLEA 232.72 USD InStock
BT219075-500ML BT219075-100ML BT219075-50ML
Zirconium, Plasma Standard Solution, Zr 1000 µg/ml, BeanTown Chemical
Standards Cationic Standards Zirconium Standards
MDL No: MFCD00011303
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty