Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium hydroxide octahydrate ≥97%

Supplier: BeanTown Chemical

220955-2.5KG 220955-500G 220955-10KG
BT220955-10KGEA 414.52 USD InStock
BT220955-10KG BT220955-2.5KG BT220955-500G
Barium hydroxide octahydrate ≥97%
Barium hydroxide octahydrate
  • RTECS: CQ9200000
  • Air Sensitive, Hygroscopic
  • Solid


Formula: Ba(OH)₂·8H₂O
MW: 315.47 g/mol
Melting Pt: 78 °C
MDL Number: MFCD00149152
CAS Number: 12230-71-6
EINECS: 241-234-5
UN: 3262
ADR: 8,III
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty