Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Pentane ≥99.0% for spectrophotometry

Supplier: BeanTown Chemical

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Synonyms: Pentane

224215-1L 224215-4X1L 224215-4L
BT224215-4X1LEA 155.8 USD InStock
BT224215-4X1L BT224215-4L BT224215-1L
n-Pentane ≥99.0% for spectrophotometry
n-Pentane
  • RTECS: RZ9450000
  • Refractive index: 1.358
  • Liquid


Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 36 °C
Melting Pt: –130 °C
Density: 0.626 g/mL
Flash Pt: –49 °C (–56 °F)
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/81/67/738167.jpg
Order Now
Specification Test Results
Assay 99.0% min.

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty