Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroform ≥99.8% stabilized ACS for HPLC

Supplier: BeanTown Chemical

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Synonyms: Methane, trichloro-TrichloroformTrichloromethane

Stabilization: Stabilised with ethanol

224225-4L 224225-4X4L 224225-1L
BT224225-1LEA 55.56 USD InStock
BT224225-1L BT224225-4L BT224225-4X4L
Chloroform ≥99.8% stabilized ACS for HPLC
Chloroform
  • RTECS: FS9100000
  • Refractive index: 1.445
  • Liquid


Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 60.5…61.5 °C
Melting Pt: –63 °C
Density: 1.492 g/mL
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/69/36/726936.jpg
Order Now
Specification Test Results
Assay 99.8% min.
Color (APHA) ≤10
Residue after evaporation ≤0.001%
Acetone and aldehyde Passes test
Acid and Chloride Passes test
Free chloride (Cl) Passes test
Lead (Pb) ≤0.05ppm
Substances darkened by H2SO4 Passes test
Suitability for use in dithizone test Passes test
Ethanol stabilizer 0.5-1.5%
Water ≤0.03%

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty