Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethylacetamide ≥99.5% for HPLC

Supplier: BeanTown Chemical
Synonyms: Acetic acid dimethylamideDimethyl Acetamide

224485-4L 224485-1L 224485-4X1L
BT224485-4LEA 276.76 USD InStock
BT224485-4L BT224485-4X1L BT224485-1L
N,N-Dimethylacetamide ≥99.5% for HPLC
N,N-Dimethylacetamide
  • RTECS: AB7700000
  • Refractive index: 1.437
  • Hygroscopic
  • Liquid


Formula: CH₃CON(CH₃)₂
MW: 87.12 g/mol
Boiling Pt: 165…166 °C
Melting Pt: –20 °C
Density: 0.937 g/mL
Flash Pt: 70 °C (158 °F)
MDL Number: MFCD00008686
CAS Number: 127-19-5
EINECS: 204-826-4
UN: 2810
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/13/2298913.jpg
Order Now
Specification Test Results
Assay 99.5% min.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty