Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 34.0 - 37.5%

Supplier: BeanTown Chemical

224570-500ML 224570-2.5L
BT224570-500MLEA 108.79 USD InStock
BT224570-500ML BT224570-2.5L
Hydrochloric acid 34.0 - 37.5%
Hydrochloric acid
  • RTECS: MW4025000
  • Liquid, single sub-boiling quartz distillation


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: >100 °C
Density: 1.19 g/mL
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Assay 34.0-37.5%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty