Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid ≥36.5% ≥99.999% (trace metals basis)

Supplier: BeanTown Chemical
Synonyms: Muriatic acidHydrochloric acid solution

225305-2.5L 225305-1L 225305-250ML
BT225305-2.5LEA 367.61 USD InStock
BT225305-2.5L BT225305-250ML BT225305-1L
Hydrochloric acid ≥36.5% ≥99.999% (trace metals basis)
Hydrochloric acid
  • RTECS: MW4025000
  • Liquid


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: >100 °C
Density: 1.19 g/mL
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Assay 99.999%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty