Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Certified Reference Materials, Sulphide, Rocklabs
Supplier: ROCKLABS
SE-AU-100X100G SG-AU-100X30G SL-AU-2.5KG SJ-AU-100X50G SL-AU-100X30G SI-AU-100X100G SP-AU-2.5KG SH-AU-100X100G SG-AU-100X100G SF-AU-100X30G SE-AU-100X50G SK-AU-2.5KG SI-AU-100X30G SI-AU-2.5KG SQ-AU-2.5KG SN-AU-2.5KG SQ-AU-100X30G SG-AU-2.5KG SJ-AU-100X100G SN-AU-100X30G SK-AU-100X30G SJ-AU-100X30G SH-AU-100X50G SP-AU-100X30G SP-AU-100X100G SH-AU-100X30G SN-AU-100X50G SK-AU-100X100G SH-AU-2.5KG SF-AU-2.5KG SG-AU-100X50G SI-AU-100X50G SF-AU-100X50G SK-AU-100X50G SN-AU-100X100G SL-AU-100X100G SL-AU-100X50G SQ-AU-100X50G SE-AU-2.5KG SP-AU-100X50G SF-AU-100X100G SJ-AU-2.5KG SE-AU-100X30G SQ-AUAG-2.5KG SQ-AU-100X100G
CA10032-626EA 276 USD InStock
CA10067-800 CA10067-822 CA10067-866 CA10067-844 CA10067-920 CA10067-886 CA10067-862 CA10032-626 CA10067-882 CA10067-860 CA10032-602 CA10067-818 CA10067-916 CA10067-816 CA11024-112 CA10067-912 CA10067-878 CA10067-812 CA10067-856 CA10067-834 CA10032-654 CA10067-932 CA10067-832 CA10032-652 CA10067-874 CA10067-852 CA10032-636 CA10067-894 CA10067-870 CA10067-890 CA10029-434 CA10067-790 CA10029-282 CA10029-284 CA10067-908 CA10067-808 CA10067-928 CA10067-828 CA10067-904 CA10032-628 CA10032-606 CA10067-848 CA10067-826 CA10067-804 CA10067-924
Certified Reference Materials, Sulphide, Rocklabs
Standards Environmental Analysis Standards
Rocklabs Gold Standards are reference materials that are certified for either Pt, Pd, Au or Ag
Recommended Gold (ppm) concentrations. Packaged in jars or satchets.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty