Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium silicate ∼41° Baume in aqueous solution

Supplier: VWR
Danger

ANAC85054-540
CA11021-796EA 29.94 USD InStock
CA11021-796
Sodium silicate ∼41° Baume in aqueous solution
Sodium silicate


Formula: Na₂O(SiO₂)x·xH₂O
MDL Number: MFCD00003492
CAS Number: 1344-09-8
EINECS: 215-687-4
UN: 3266
ADR: 8,III
Order Now
Specification Test Results
Sodium monoxide (Na2O) 8.70 - 9.10%
Silica (SiO2) 28.20 - 29.00%
Ratio (SiO2/Na2O) 3.15 - 3.27%
Solids 36.90 - 38.10 %
Baumé at 20°C 40.5 - 41.5

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty