Designed for coal and coke analysis procedures.
- Enlarged top vent connects easily to exhaust systems for ventilation of contaminants
- Heating elements can be replaced individually, cutting down on operational cost
Furnaces come with an adjustable gas flow meter/valve (0–80L/min.) located on the front for easy adjustment of airflow rate
Choose from two furnace sizes. Available with either 4- or 16-segment programmable controller. Type 6000 furnace meets ASTM D3174.
Stainless steel manifold at rear chamber pre-warms incoming gases providing maximum temperature gradient of only ±3°C at 750°C (1382°F). Heating elements are enveloped in a custom formed refractory cement, reducing the damage caused by contaminant build-up on the resistant wire.
Certifications: CSA certified.
Ordering information: Furnaces include a 3-wire cord with plug. Hearth plate and replacement heating elements available separately.