Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium nitrate ≥99.0%, crystals ACS

Supplier: VWR
Warning

ANAC83720-300
CA71008-746EA 150.98 USD InStock
CA71008-746
Sodium nitrate ≥99.0%, crystals ACS
Sodium nitrate


Formula: NaNO₃
MW: 84.99 g/mol
Boiling Pt: 380 °C (1013 hPa)
Melting Pt: 308 °C
Density: 2.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011119
CAS Number: 7631-99-4
EINECS: 231-554-3
UN: 1498
ADR: 5.1,III
REACH: 01-2119488221-41
Merck Index: 12,08792
Order Now
Specification Test Results
Assay (NaNO3) 99.0% min.
pH of a 5% Solution at 25°C 5.5 - 8.3
Insoluble Matter 0.005% max.
Chloride (Cl) 0.001% max.
Iodate (IO3) 5 ppm max.
Nitrite (NO2) 0.001% max.
Phosphate (PO4) 5 ppm max.
Sulfate (SO4) 0.003% max.
Calcium (Ca) 0.005% max.
Magnesium (Mg) 0.002% max.
Heavy metals (as Pb) 5 ppm max.
Iron (Fe) 3 ppm max.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty