Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Toluene, Accusolv™, distilled

Supplier: Avantor
Danger
Synonyms: MethacideBenzene, methylMethylbenzolToluolPhenylmethaneMethylbenzeneMethane, phenyl-

ANAC92362-540
CA71009-106CS 1053.22 USD InStock
CA71009-106
Toluene, Accusolv™, distilled
Toluene


Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 12,09667
https://us-legacy.vwr.com/stibo/web/std.lang.all/61/73/766173.jpg
Order Now
Specification Test Results
UV absorbance (1 cm cell vs H2O) 285 nm 1.000 max.
UV absorbance (1 cm cell vs H2O) 300nm 0.120 max.
UV absorbance (1 cm cell vs H2O) 325nm 0.020 max.
UV absorbance (1 cm cell vs H2O) 350nm 0.005 max.
UV absorbance (1 cm cell vs H2O) 400nm 0.005 max.
Assay(GC) 99.8% min.
Water(H2O) 0.03% max.
Residue after evaporation 1 mg/L max.
Benzene 500 mg/L max.
Electron capture GC as heptachlorepoxide 10 ng/L max.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty