Would you like to log in again?
Sample Tubes, 50 ml, Simport Scientific
Supplier: Simport
C571-1 C571-2 C571-2
CA73960-336CS 874.25 USD InStock
CA73960-336 CA15705-376 14231-882
Sample Tubes, 50 ml, Simport Scientific
Tubes Test Tubes
These polypropylene, conical bottom tubes with self-standing bases have molded-in graduations from 10 to 50 ml in 5 ml increments.
  • Chemical-resistant and shatterproof
  • With a leakproof screw cap, which is particularly important for transporting hazardous material
Ideal for transport and storage of urine, sputum, and most liquids or particulate samples.
Temperature: –90 to 121 °C (–130 to 250 °F).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty