Would you like to log in again?
Batch Pressure Filters, Floor Mounted, Sepor
Supplier: Sepor
070B-055 070B-001 070B-056 070B-003
CA76047-094EA 15944.23 USD InStock
CA76047-094 CA76047-092 CA76047-090 CA76047-088
Batch Pressure Filters, Floor Mounted, Sepor
Filters Membrane Filter Units
Floor Mounted Batch Pressure Filters are manufactured for batch filtering of slurries under pressure, up to 75 psi. They offer a quick, convenient method of dewatering slurries, in two sizes, 11.5 gallon and 3 gallon capacity.
Units feature a screw jack lid opening and closing mechanism, swing away design for filling, inspection or cleaning
Also included with each pressure filter is a filter cloth, a package of 100 high wet strength filter papers with a 25 nominal micron pore opening, internal piping with pressure relief valve, pressure gauge, bleed off valve, and drain valve. All that is required for operation is a slurry, and a source of compressed air (at least 1 CFM @ 60 psi).
The base plate with the filter grid has the same screw jack opening and closing mechanism and swing away design for easy removal of the filtered solids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty