Would you like to log in again?
EVERHUTCH NESTCART Mobile Utility Carts, Kewaunee
Supplier: Kewaunee
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MCB00D3030M-01 MCB40E3024LM1 MCB50D3324M-01 MCB00D3018M-01 MCB30D3318M-00 MCB50D3618M-00 MCB50D3324M-00 MCB30D3324M-00 MCB00D3630M-00 MCB03C3624LM0 MCB00D3630M-01 MCB30D3024M-01 MCB50D3618M-01 MCB40E3024LM0 MCB00D3318M-00 MCB50D3624M-00 MCB00D3624M-01 MCB26D3018M-01 MCB30D3030M-00 MCB32D3330M-00 MCB00D3324M-01 MCB00D3018M-00 MCB50D3318M-00 MCB51D3630M-01 MCB03C3624R1 MCB30D3030M-01 MCB52D3630M-01 MCB08D3624M-00 MCB03C3624RM0 MCB00D3624M-00 MCB03C3624LM1 MCB30D3018M-01 MCB08D3618M-00 MCB00D3030M-00 MCB50D3318M-01 MCB30D3024M-00 MCB30D3630M-00 MCB30D3324M-01 MCB00D3318M-01 MCB08D3618M-01 MCB00D3618M-01 MCB00D3024M-01 MCB00D3618M-00 MCB30D3318M-01 MCB00D3324M-00 MCB30D3018M-00 MCB50D3624M-01 MCB26D3018M-00 MCB00D3024M-00 MCB52D3630M-00 MCB32D3330M-01 MCB08D3624M-01 MCB51D3630M-00
CB00D303001EA 2810 USD InStock
CB00D303001 CB30D301800 CB50D362400 CB30D301801 CB00D332401 CB00D361801 CB00D332400 CB08D361801 CB08D361800 CB00D361800 CB50D362401 CB00D302401 CB00D302400 CB52D363001 CB52D363000 CB30D331801 CB30D363000 CB50D332400 CB50D332401 CB30D331800 CB40E3024L01 CB40E3024L00 CB00D301800 CB00D301801 CB30D303001 CB30D303000 CB00D363001 CB50D331801 CB30D332401 CB30D332400 CB00D363000 CB50D331800 CB26D301800 CB26D301801 CB03C3624L01 CB03C3624L00 CB51D363001 CB00D331801 CB30D302401 CB00D331800 CB51D363000 CB30D302400 CB03C3624R01 CB03C3624R00 CB50D361800 CB32D333001 CB50D361801 CB00D362400 CB08D362401 CB00D362401 CB08D362400 CB00D303000 CB32D333000
EVERHUTCH NESTCART Mobile Utility Carts, Kewaunee
Carts
Mobile units with drawer configurations for storage in clinical and work areas.
  • Mobile cart on wheels
  • Sitting, ADA, and standing height cabinets
  • Drawer configurations
  • Under counter storage
  • File cabinet
  • Supply storage
Ideal for supply storage in clinical areas such as patient rooms. Provides added storage space under a counter to maximize the space. Variable heights allow for sitting, ADA, and standing height work surfaces (30, 33, 36") with full extension drawers. The cart is on four swivel locking casters for easy movement.
Ordering information: The 00 product indicates no locks and 01 indicates all drawers will have a key lock installed.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty