Would you like to log in again?
VWR® Contour® Lock Rails
Supplier: VWR
VWR® Contour™
CF3-0133-A CF3-0121-00 CF3-0115-A CF3-0145-A CF3-0115-00 CF3-0115PB-A CF3-0121-A
CF3-0115-00EA 72.99 USD InStock
CF3-0115-00 CF3-0115-A CF3-0133-A CF3-0121-A CF3-0145-A CF3-0121-00 CF3-0115PB-A
VWR® Contour® Lock Rails
Furniture Casework
Lock rails for 18 to 48" VWR® base units.
  • Use with base cabinet door locks
  • Required on all drawers except top drawers of cabinet
  • For use with half-width drawers
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty