Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 25, 2024. During this time, our site will be not available. We appreciate your patience.
VWR® Contour™ Top-Mounted Apron Legs
Supplier: VWR
VWR® Contour™
CWA-1159-A CFA-2159-A CWA-2159-A CFA-1159-A
CWA-1159-AEA 142.96 USD InStock
CWA-1159-A CFA-2159-A CWA-2159-A CFA-1159-A
VWR® Contour™ Top-Mounted Apron Legs
Furniture Casework
Each 5.1 cm (2") square leg is welded to 10.2 cm (4") square steel plate.
  • Contour casework is manufactured of prime grade, cold-rolled, U.S. Standard sheet steel
  • No ragged edges
  • Light neutral color
The complete apron is used to create a knee space for a desk area or to create a free-standing table. Apron components are sold separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty