Would you like to log in again?
VWR® Contour™ Apron Rollout Keyboard Shelves
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR® Contour™
VWR0221 VWR0220 CWA01M0219-03 CWA01M0220-03 VWR0219 CWA01M0221-03
CWA01M0219-03EA 731.81 USD InStock
CWA01M0219-03 CWA01M0220-03 CWA01M0221-03 CFA01M0219-03 CFA01M0221-03 CFA01M0220-03
VWR® Contour™ Apron Rollout Keyboard Shelves
Furniture Casework
Rollout keyboard shelves feature a low-profile design and come in a range of lengths.
  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
  • Color: light neutral
These shelves feature smooth surfaces with coatings to resist against chemicals.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty