Would you like to log in again?
VWR® Contour™ Sitting Height Base Cabinets, Cupboard Cabinets
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR® Contour™
CWC-1032-A CWC-1034-L CFC-1032-A CFC-1034-12 CWC-1038-A CFC-1034-A CFC-1032-12 CFC-1038-A CWC-1036-A CFC-1036-A C03M3018L03 CFC-1032-L CWC-1034-A CWC-1032-L CFC-1036-12 CFC-1034-L
CWC-1032-LEA 1490.26 USD InStock
CWC-1032-L CFC-1032-12L CFC-1034-L CFC-1032-L CWC-1034-L CWC-1034-A CWC-1032-A CFC-1036-A CWC-1038-A CFC-1034-A CFC-1032-12 CFC-1034-12 CWC-1036-A CFC-1032-A CFC-1036-12 CFC-1038-A
VWR® Contour™ Sitting Height Base Cabinets, Cupboard Cabinets
Furniture Casework
Heavy-gauge steel full-frame construction.
  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
  • Light neutral color
A 2.54 cm (1") thick work surface creates an overall height of 76.2 cm (30"). Cupboard cabinets are supplied with one full-width adjustable shelf.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty