Would you like to log in again?
VWR® Contour™ Sitting Height Base Cabinet, Drawer Cabinets
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR® Contour™
CFD-1302-10 CFD-1304-A CWD-1304-A CFD-1842-10 CFD-1262-10 CWD-1812-A CFD-1302-A CFD-1262-A CFD-1842-A CFD-1812-10 CWD-1302-A CWD-1842-A CWD-1262-A CFD-1304-10 CFD-1812-A
CFD-1812-10EA 1481.64 USD InStock
CFD-1812-10 CFD-1262-10 CFD-1842-A CFD-1812-A CWD-1302-A CWD-1304-A CFD-1262-A CWD-1262-A CWD-1812-A CFD-1302-10 CWD-1842-A CFD-1304-A CFD-1842-10 CFD-1304-10 CFD-1302-A
VWR® Contour™ Sitting Height Base Cabinet, Drawer Cabinets
Furniture Casework
Heavy-gauge steel full-frame construction.
  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
A 2.54 cm (1”) thick work surface creates an overall height of 76.2 cm (30”). All cupboard cabinets are supplied with one full-width adjustable shelf.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty